Nickel(II) sulphamate tetrahydrate, Sigma-Aldrich®
Catalog # SIAL262277-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: Nickel (II) sulphamidate tetrahydrate, Nickel bis(sulphamidate) tetrahydrate, Nickel sulphamate tetrahydrate, Nickel(II)sulphamate tetrahydrate
:
:322.92 g/mol
EINECS:237-396-1
UN:1259
:GHS08
:GHS09
CAS Number:124594-15-6
Formula:H₄N₂NiO₆S₂·4H₂O
MDL Number:MFCD00149966
ADR:6.1 I
:M-Clarity™ quality level = MQ100