L(+)-Lysine monohydrochloride
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:657-27-2
:
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MDL Number:MFCD00064564
Merck Index:13,05656
Boiling Pt:290 °C (1013 hPa)
Melting Pt:263 °C (lit.)
:182.65 g/mol
Storage Temperature:Ambient
EINECS:200-294-2
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- pH (10% aq soln):5-6
- Loss on Drying:≤1.0%
- Optical Rotation:+20° to +23° (c=8, 2N HCl)
- Heavy Metals:≤15ppm
Specification Test Results
Frequently Bought Together