L(+)-Lysine monohydrochloride, Sigma-Aldrich®
: Merck
:657-27-2
:
:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
:MFCD00064564
:263…264 °C
:M-Clarity™ quality level = MQ200
:182.65 g/mol
:Ambient
:211-519-9
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Iron(II) sulphate heptahydrate 99+% ACS
: Thermo Fisher Scientific