Order Entry
Austria
ContactUsLinkComponent
L(+)-Lysine monohydrochloride for synthesis, Sigma-Aldrich®
L(+)-Lysine monohydrochloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
L(+)-Lysine monohydrochloride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    420 g/l (20 °C)
  • Melting Point:
    263 - 264 °C
  • Molar Mass:
    182.64 g/mol
  • Bulk Density:
    360 kg/m3
  • pH:
    5.5 - 6.0 (100 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together