Order Entry
Austria
ContactUsLinkComponent
Sodium diethyldithiocarbamate trihydrate ACS analytical reagent, Supelco®
Irritant
Environment
Sodium diethyldithiocarbamate trihydrate ACS analytical reagent, Supelco®
Supplier:  Merck
Certificates
Irritant
Environment
Sodium diethyldithiocarbamate trihydrate ACS analytical reagent, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    600 g/l (20 °C)
  • Melting Point:
    93 °C
  • Molar Mass:
    225.31 g/mol
  • pH:
    7 (20 g/l, H2O, 20 °C)
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous hazardous material

Specification Test Results

Related Information

Frequently Bought Together