Order Entry
Austria
ContactUsLinkComponent
 
Magnesium sulphate 1.0 M MgSO4 volumetric solution, Fluka™
Catalog # 63126-1L
CAS Number:  
undefined
Magnesium sulphate 1.0 M MgSO4 volumetric solution, Fluka™
Catalog # 63126-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Factor:
    0.995 - 1.005

Specification Test Results

About this item

Synonyms: MgSO4, Magnesium sulphate anhydrous
 :
 :120.37 g/mol
Boiling Pt:100 °C
Density:1.10 g/cm³
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110