
Barium chloride 0.1 M BaCl2 (0.2N) volumetric solution, Fluka™
Catalog # 34252-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:208.24 g/mol
Density:1.00 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-788-1
ADR:6.1,III
:GHS07
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445