Order Entry
Austria
ContactUsLinkComponent
Sodium chloride 5 mol/l (5 N) in aqueous solution for biotechnology
Sodium chloride 5 mol/l (5 N) in aqueous solution for biotechnology
Catalog # E529-500ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium chloride 5 mol/l (5 N) in aqueous solution for biotechnology
Catalog # E529-500ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Conductivity (1 %, Water):
    5000 - 7000 umhos
  • Identification:
    PASS
  • pH @25 °C:
    5.0 - 9.0

Specification Test Results

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
CAS Number:7647-14-5
 :
Formula:NaCl
MDL Number:MFCD00003477
 :58.44 g/mol
Boiling Pt:1413
Melting Pt:801
Storage Temperature:Ambient
EINECS:231-598-3
Merck Index:14,08599