Order Entry
Austria
ContactUsLinkComponent
Sodium chloride 5 mol/l (5 N) in aqueous solution
Sodium chloride 5 mol/l (5 N) in aqueous solution
Catalog # AMBI9760G
Supplier:  Ambion
CAS Number:  
Sodium chloride 5 mol/l (5 N) in aqueous solution
Catalog # AMBI9760G
Supplier:  Ambion
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
Boiling Pt:1413
Melting Pt:801
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477
Merck Index:14,08599