Organic reference standard, 2-Nitrophenol 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA15590200ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:88-75-5
- Description:2-Nitrophenol 100 µg/ml in methanol
- Formula:C₆H₅NO₃
- Description:2-Nitrophenol