Order Entry
Austria
ContactUsLinkComponent
VWR® ARISTAR®, Tin Standard Solution, 10000 mg/L Sn in dil. Nitric Acid with Hydrofluoric Acid (max. 1%) (from Sn), Standard for ICP
VWR® ARISTAR®, Tin Standard Solution, 10000 mg/L Sn in dil. Nitric Acid with Hydrofluoric Acid (max. 1%) (from Sn), Standard for ICP
Catalog # 456232S
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Tin Standard Solution, 10000 mg/L Sn in dil. Nitric Acid with Hydrofluoric Acid (max. 1%) (from Sn), Standard for ICP
Catalog # 456232S
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Tin
  • Symbol:
    Sn

Specifications

About this item

Sn in HNO₃ tr. HF.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities

Supplied with certificate of analysis.