Ytterbium standard solution, 1000 mg/L Yb in 5% nitric acid (from Yb₂O₃), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular formula: Yb₂O₃ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011286
Certifications: Traceable to NIST.
Specifications
- MDL No.:MFCD00011286
Specifications
Related Information
Frequently Bought Together