Order Entry
Austria
ContactUsLinkComponent
VWR® ARISTAR®, Thallium Standard Solution, 1000 mg/L Tl in dil. Nitric Acid (from Tl), Standard for ICP
VWR® ARISTAR®, Thallium Standard Solution, 1000 mg/L Tl in dil. Nitric Acid (from Tl), Standard for ICP
Catalog # 456162V
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Thallium Standard Solution, 1000 mg/L Tl in dil. Nitric Acid (from Tl), Standard for ICP
Catalog # 456162V
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Thallium
  • Symbol:
    Tl

Specifications

About this item

Tl in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.