Terbium standard solution, 10000 mg/L Tb in 5% nitric acid (from Tb₄O₇), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular formula: Tb₄O₇ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011256
Traceable to NIST
Certifications: Traceable to NIST.
Specifications
- MDL No.:MFCD00011256
Specifications
Related Information
Frequently Bought Together