VWR® ARISTAR®, Terbium Standard Solution, 1000 mg/L Tb in dil. Nitric Acid (from Tb₄O₇), Standard for ICP
Lieferant: VWR Chemicals
Zertifikate
Über diesen Artikel
Tb₄O₇ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Zugehörige Informationen
Zertifikate
Suche nach Chargennummer125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125) 500 ml Plastic bottle for liquids DIN 28 (ZPACK15150500)
Verpackungsinformationen
sds_VWRC45614
Sicherheitsdatenblatt (SDS)
VWR® for Calibrations VWR® Trace Analysis.pdf
Produktliteratur
Frequently Bought Together