Order Entry
Austria
ContactUsLinkComponent
Terbium standard solution, 10 mg/L Tb in 5% nitric acid (from Tb₄O₇), standard for ICP, Specpure®
Terbium standard solution, 10 mg/L Tb in 5% nitric acid (from Tb₄O₇), standard for ICP, Specpure®
Catalog # 45266.AE
CAS Number:  
Terbium standard solution, 10 mg/L Tb in 5% nitric acid (from Tb₄O₇), standard for ICP, Specpure®
Catalog # 45266.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Terbium
  • Symbol:
    Tb

Specifications

About this item

Certifications: Traceable to NIST.