
Potassium permanganate concentrate for dilution to 1L, 1/50 M KMnO4 (0.1N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
ADR:9,III
:GHS05
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together