Silicon standard solution, 10000 mg/L Si in 4% sodium hydroxide solution (from SiO₂) Certipur® standard for ICP, Supelco®
Catalog # 1.70386.0100
Supplier: Merck
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:4% NaOH
- Pack type:Plastic bottle
- Pk:100 mL
- Environmentally Preferable:
- M-Clarity quality level:MQ100
- Element:Silicon
- Symbol:Si
Specifications
About this item
Certifications: Traceable to SRM from NIST.