Order Entry
Austria
ContactUsLinkComponent
Potassium permanganate concentrate for dilution to 1L, 1/500 M KMnO4 (0.01N) volumetric solution, Fluka™
Environment
Potassium permanganate concentrate for dilution to 1L, 1/500 M KMnO4 (0.01N) volumetric solution, Fluka™
Catalog # 38136-1EA
CAS Number:  
Environment
Potassium permanganate concentrate for dilution to 1L, 1/500 M KMnO4 (0.01N) volumetric solution, Fluka™
Catalog # 38136-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
ADR:9,III
 :GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364