Order Entry
Austria
ContactUsLinkComponent
VWR®, Rubidium standard solution, 10000 mg/L Rb in dil. nitric acid (from RbNO3) ARISTAR® standard for ICP
VWR®, Rubidium standard solution, 10000 mg/L Rb in dil. nitric acid (from RbNO3) ARISTAR® standard for ICP
Catalog # 455912Y
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Rubidium standard solution, 10000 mg/L Rb in dil. nitric acid (from RbNO3) ARISTAR® standard for ICP
Catalog # 455912Y
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Rubidium
  • Symbol:
    Rb

Specifications

About this item

RbNO₃ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.