Order Entry
Austria
ContactUsLinkComponent
Mercury standard solution, 1000 mg/L Hg in 10% nitric acid (from Hg(NO₃)₂ Certipur® standard for ICP, Supelco®
Mercury standard solution, 1000 mg/L Hg in 10% nitric acid (from Hg(NO₃)₂ Certipur® standard for ICP, Supelco®
Catalog # 1.70333.0100
Supplier:  Merck
CAS Number:  
undefined
Mercury standard solution, 1000 mg/L Hg in 10% nitric acid (from Hg(NO₃)₂ Certipur® standard for ICP, Supelco®
Catalog # 1.70333.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item

Certifications: Traceable to SRM from NIST.