VWR® ARISTAR®, Praseodymium Standard Solution, 1000 mg/l Pr in dil. Nitric Acid (from Pr6O11), Standard for ICP
Lieferant: VWR Chemicals
Zertifikate
Über diesen Artikel
Pr₆O₁₁ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Zugehörige Informationen
Zertifikate
Suche nach Chargennummer125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125)
Verpackungsinformationen
sds_VWRC45584
Sicherheitsdatenblatt (SDS)
VWR® for Calibrations
Produktliteratur
Frequently Bought Together