Nickel standard solution, 1000 mg/L Ni in dil. nitric acid (from Ni(NO₃)₂) Certipur® standard for ICP, Supelco®
1.70336.0100
: Merck
:
- Concentration:1000 mg/L
- Matrix:2-3% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- M-Clarity quality level:MQ100
- Element:Nickel
- Symbol:Ni
: Traceable to SRM from NIST.