Order Entry
Austria
ContactUsLinkComponent
 
Sodium standard solution, 10000 mg/L Na in dil. nitric acid (from NaNO₃) Certipur® standard for ICP, Supelco®
  1.70381.0100
 :  Merck
 :  
Sodium standard solution, 10000 mg/L Na in dil. nitric acid (from NaNO₃) Certipur® standard for ICP, Supelco®
  1.70381.0100
 :  Merck
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Sodium
  • Symbol:
    Na

 

 

 : Traceable to SRM from NIST.