VWR® AVS TITRINORM, Molybdenum Standard Solution, 1000 mg/L Mo in Water, Standard for AAS
Supplier: VWR Chemicals
Certificates
About this item
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together