Order Entry
Austria
ContactUsLinkComponent
 
VWR® AVS TITRINORM, Lithium Standard Solution, 1000 mg/L Li in dil. Nitric Acid AVS TITRINORM Standard for AAS
Catalog # 86688.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® AVS TITRINORM, Lithium Standard Solution, 1000 mg/L Li in dil. Nitric Acid AVS TITRINORM Standard for AAS
Catalog # 86688.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item

  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%