Order Entry
Austria
ContactUsLinkComponent
VWR® ARISTAR®, Lithium Standard Solution, 1000 mg/L Li, Standard for Ion Chromatography
VWR® ARISTAR®, Lithium Standard Solution, 1000 mg/L Li, Standard for Ion Chromatography
Artikelnr. 458242F
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Lithium Standard Solution, 1000 mg/L Li, Standard for Ion Chromatography
Artikelnr. 458242F
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O/tr HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item

Li in H₂O tr.HNO₃.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.