Order Entry
Austria
ContactUsLinkComponent
VWR®, Lanthanum standard solution, 10000 mg/L La in dil. nitric acid (from La₂O₃) ARISTAR® standard for ICP
VWR®, Lanthanum standard solution, 10000 mg/L La in dil. nitric acid (from La₂O₃) ARISTAR® standard for ICP
Catalog # 455554G
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Lanthanum standard solution, 10000 mg/L La in dil. nitric acid (from La₂O₃) ARISTAR® standard for ICP
Catalog # 455554G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Lanthanum
  • Symbol:
    La

Specifications

About this item

La₂O₃ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.