Order Entry
Austria
ContactUsLinkComponent
 
Lanthanum standard solution, 1000 mg/L La in dil. nitric acid (from La(NO₃)₃) Certipur® standard for ICP, Supelco®
Catalog # 1.70327.0100
Supplier:  Merck
CAS Number:  
undefined
Lanthanum standard solution, 1000 mg/L La in dil. nitric acid (from La(NO₃)₃) Certipur® standard for ICP, Supelco®
Catalog # 1.70327.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Lanthanum
  • Symbol:
    La

Specifications

About this item

Certifications: Traceable to SRM from NIST.