VWR® ARISTAR®, Copper Standard Solution, 10000 mg/L Cu in dil. Nitric Acid (from Cu), Standard for ICP
Supplier: VWR Chemicals
Certificates
About this item
Cu in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Related Information
Certificates
Search by Lot Number125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125) 500 ml Plastic bottle for liquids DIN 28 (ZPACK15150500)
Packaging Information
VWR® for calibration VWR® for Calibrations VWR® for Calibration
Product Literature
Frequently Bought Together