Copper standard solution, 1000 mg/L Cu in dilute nitric acid (from Cu(NO₃)₂) Certipur® standard for ICP, Supelco®
1.70314.0100
: Merck
:
- Concentration:1000 mg/L
- Matrix:2-3% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- M-Clarity quality level:MQ100
- Element:Copper
- Symbol:Cu
: Traceable to SRM from NIST.