Copper standard solution, 1000 mg/L Cu in 0,5 M nitric acid (from Cu(NO₃)₂) Certipur® standard for AAS, Supelco®
Supplier: Merck
Certificates
About this item
Certifications: Traceable to SRM from NIST.
Specifications
- M-Clarity quality level:MQ100
Specifications
Related Information
Certificates
Search by Lot NumberCertipur
Product Literature
msds_119786
SDS
ProductPage_119786
Product specification
Frequently Bought Together