Order Entry
Austria
ContactUsLinkComponent
 
Potassium standard solution, 1000 mg/L K in dilute nitric acid (from KNO₃) Certipur® standard for ICP, Supelco®
  1.70342.0100
 :  Merck
 :  
undefined
Potassium standard solution, 1000 mg/L K in dilute nitric acid (from KNO₃) Certipur® standard for ICP, Supelco®
  1.70342.0100
 :  Merck
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2-3% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Potassium
  • Symbol:
    K

 

 

 : Traceable to SRM from NIST.