ARISTAR®VWR® , Potassium Standard Solution, 200 mg/L K in dilute Nitric Acid (from K), Standard for Ion Chromatography
Artikelnr. 458302A
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:200 mg/L
- Matrix:H₂O/tr HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Potassium
- Symbol:K
Specifications
About this item
K in H₂O tr.HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.