Order Entry
Austria
ContactUsLinkComponent
 
Iridium standard solution, 1000 mg/L Ir in 7% hydrochloric acid (from IrCl₃) Certipur® standard for ICP, Supelco®
  1.70325.0100
 :  Merck
 :  
undefined
Iridium standard solution, 1000 mg/L Ir in 7% hydrochloric acid (from IrCl₃) Certipur® standard for ICP, Supelco®
  1.70325.0100
 :  Merck
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    7% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Iridium
  • Symbol:
    Ir

 

 

 : Traceable to SRM from NIST.