Indium standard solution, 1000 mg/L In in dilute nitric acid (from In(NO?)?) Certipur® standard for ICP, Supelco®
Catalog # 1.70324.0100
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- M-Clarity quality level:MQ100
- Element:Indium
- Symbol:In
Specifications
About this item
Certifications: Traceable to SRM from NIST.