Order Entry
Austria
ContactUsLinkComponent
ARISTAR®VWR® , Erbium Standard Solution, 10000 mg/l Er in dil. Nitric Acid (from Er2O3), Standard for ICP
ARISTAR®VWR® , Erbium Standard Solution, 10000 mg/l Er in dil. Nitric Acid (from Er2O3), Standard for ICP
Catalog # 455334T
Supplier:  VWR Chemicals
CAS Number:  
undefined
ARISTAR®VWR® , Erbium Standard Solution, 10000 mg/l Er in dil. Nitric Acid (from Er2O3), Standard for ICP
Catalog # 455334T
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Erbium
  • Symbol:
    Er

Specifications

About this item

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Supplied with certificate of analysis.