Order Entry
Austria
ContactUsLinkComponent
 
ARISTAR®VWR® , Cerium Standard Solution, 10000 mg/L Ce in dil. Nitric Acid (from (NH₄)₂Ce(NO₃)₆), Standard for ICP
  455192A
 :  
undefined
ARISTAR®VWR® , Cerium Standard Solution, 10000 mg/L Ce in dil. Nitric Acid (from (NH₄)₂Ce(NO₃)₆), Standard for ICP
  455192A
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Cerium
  • Symbol:
    Ce

 

 

(NH₄)₂ Ce(NO₃)₆ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.