Order Entry
Austria
ContactUsLinkComponent
Boron standard solution, 1000 mg/L B in water (from B(OH₃)) Certipur® standard for ICP, Supelco®
Boron standard solution, 1000 mg/L B in water (from B(OH₃)) Certipur® standard for ICP, Supelco®
Catalog # 1.70307.0100
Supplier:  Merck
CAS Number:  
undefined
Boron standard solution, 1000 mg/L B in water (from B(OH₃)) Certipur® standard for ICP, Supelco®
Catalog # 1.70307.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item

H₃BO₃ in water.

Certifications: Traceable to SRM from NIST. The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Delivery: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.