Order Entry
Austria
ContactUsLinkComponent
 
VWR® ARISTAR®, Beryllium Standard Solution, 10000 mg/L Be in dil. Nitric Acid, Standard for ICP
  455092U
 :  
undefined
VWR® ARISTAR®, Beryllium Standard Solution, 10000 mg/L Be in dil. Nitric Acid, Standard for ICP
  455092U
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Beryllium
  • Symbol:
    Be

 

 

BeO.(Be(OAc)₂)₃) in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.