Order Entry
Austria
ContactUsLinkComponent
Sulphate standard solution, 1000 mg/L SO₄²⁻ in Water (from Na₂SO₄) Certipur®, Supelco®
Sulphate standard solution, 1000 mg/L SO₄²⁻ in Water (from Na₂SO₄) Certipur®, Supelco®
Catalog # 1.19813.0500
Supplier:  Merck
CAS Number:  
Sulphate standard solution, 1000 mg/L SO₄²⁻ in Water (from Na₂SO₄) Certipur®, Supelco®
Catalog # 1.19813.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Sulphate standard solution
  • M-Clarity quality level:
    MQ100
  • Contents:
    Na₂SO₄ in H₂O

Specifications

About this item