Order Entry
Austria
ContactUsLinkComponent
Chloride standard solution, 1,000 mg/L Cl- in Water (from NaCl) Certipur®, Supelco®
Chloride standard solution, 1,000 mg/L Cl- in Water (from NaCl) Certipur®, Supelco®
Catalog # 1.19897.0500
Supplier:  Merck
CAS Number:  
Chloride standard solution, 1,000 mg/L Cl- in Water (from NaCl) Certipur®, Supelco®
Catalog # 1.19897.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Chloride standard solution
  • M-Clarity quality level:
    MQ100
  • Contents:
    NaCl in H₂O

Specifications

About this item