(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-79-8
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00063238
:132.16 g/mol
Merck Index:13,05233
Boiling Pt:188 °C (1013 hPa)
Density:1.04 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:202-888-7
Frequently Bought Together