

Eisen(III)chlorid 45% (FeCl₃-Basis) in wässriger Lösung, Purum, Fluka™
Catalog # 12322-4X2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Packung:Glasflasche
- VE:10 L
- Seq:5214
Specifications
Specification Test Results
- Assay (excluding Fe):44.6 - 45.4 %
Specification Test Results
About this item
:
:162.21 g/mol
Density:1,48 g/cm3 (20 °C)
Storage Temperature:Raumtemperatur
EINECS:231-729-4
UN:2582
ADR:8,III
:GHS07
:GHS05
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005