
Barium chloride 20% in aqueous solution, Fluka™
Catalog # 35323-5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Assay:19 - 21 %
Specification Test Results
About this item
:
:208.24 g/mol
Boiling Pt:? 100 °C (1013 hPa)
Storage Temperature:Ambient
EINECS:233-788-1
ADR:6.1,III
:GHS07
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445