Sodium chloride 9 g/l in aqueous solution
Catalog # VWRS5909.5000
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477