Phenol liquified ≥ 89,0% stabilised USP, JP, Multi-Compendial, J.T.Baker®
: Avantor
:108-95-2
:
:C₆H₅OH
:MFCD00002143
:94.11 g/mol
:6.1,II
:Stabilised with citric acid
:203-632-7
:2821
:GHS06
:GHS05
:GHS08
:Ambient
- Environmentally Preferable:
- CAUTION: For Manufacturing, processing or repackaging:= 182.5 °C
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Distilling Range - Final Temperature:≤ 182.5 °C
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Clarity of Solution and Reaction:Passes Test
- USP - Nonvolatile Residue:≤ 0.05 %
- USP - Assay:≥ 89.0 %
- USP - Benzene:≤ 2.0 ppm
- JP - Identification (1):Passes Test
- JP - Identification (2):Passes Test
- JP - Boiling Point:≤ 182 °C
- JP - Clarity and Color of Solution and Acidity or Alkalinity:Passes Test
- JP - Residue on Evaporation:≤ 0.05 %
- JP - Assay:≥ 88.0 %
Frequently Bought Together