Order Entry
Austria
ContactUsLinkComponent
L(+)-Rhamnose monohydrate 99%
L(+)-Rhamnose monohydrate 99%
Certificates
undefined
L(+)-Rhamnose monohydrate 99%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Water:
    9.0 to 11.0 % (K.F.)
  • Heavy metals:
    ≤20 ppm
  • HPLC:
    ≥98.5 % (ELSD)
  • Residue after ignition:
    ≤0.2 %

Specification Test Results

Related Information

Frequently Bought Together