Lactose monohydrate, powder, EMPROVE® EXPERT Ph. Eur., BP, NF, JP for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:10039-26-6
:
Formula:C₁₂H₂₂O₁₁·1H₂O
:360.32 g/mol
Melting Pt:223 °C
Storage Temperature:Ambient
EINECS:200-559-2
Frequently Bought Together