Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Lactose monohydrate, Millipore®

Lactose monohydrate, Millipore®
Supplier:  Merck
Certificates
Lactose monohydrate, Millipore®

3 Options Available Below

Product Details & Documents
Synonyms:  Milk sugar, Lactobiose
  • CAS Number:10039-26-6
  • Formula:C₁₂H₂₂O₁₁·1H₂O
  • Molecular Weight:360.32 g/mol
  • Melting Pt:223 °C
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-559-2
Product Family Options

Product Information